missing translation for 'onlineSavingsMsg'
Learn More

3-Buten-1-ol 98.0+%, TCI America™

Catalog No. B023610ML
Change view
Click to view available options
Quantity:
10 mL
25 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B023610ML 10 mL
B023625ML 25 mL
B0236500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B023610ML Supplier TCI America Supplier No. B023610ML

Chemical Identifiers

CAS 627-27-0
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.107
MDL Number MFCD00002959
InChI Key ZSPTYLOMNJNZNG-UHFFFAOYSA-N
Synonym 3-buten-1-ol, allylcarbinol, 3-butenyl alcohol, 1-buten-4-ol, vinylethyl alcohol, 3-butene-1-ol, ch2=chch2ch2oh, homoallyl alcohol, unii-3db2krm1i9, 1-hydroxy-3-butene
PubChem CID 69389
IUPAC Name but-3-en-1-ol
SMILES C=CCCO

Specifications

Color Colorless
Boiling Point 114°C
Quantity 10 mL
UN Number 1987
Formula Weight 72.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Buten-1-ol
EINECSNumber (2)-4017&(2)-4081
TSCA Yes