accessibility menu, dialog, popup

UserName

CAS RN 627-27-0

IUPAC Name: but-3-en-1-ol
Synonyms: 3-buten-1-ol
Molecular Weight (g/mol): 72.11
Molecular Formula: C4H8O
InChi Key: ZSPTYLOMNJNZNG-UHFFFAOYSA-N
SMILES: OCCC=C
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)