missing translation for 'onlineSavingsMsg'
Learn More

3-Buten-1-ol, 98+%

CAS: 627-27-0 | C4H8O | 72.107 g/mol

Supplier:  Thermo Scientific Chemicals L0076706

 View more versions of this product

Catalog No. AAL0076706


Only null left
Explore available promotions

Description

Description

3-Buten-1-ol is employed in a study of the Mn-catalyzed hydrohydrazination of olefins. Also used in a study of the conversion of propargylic acetates to ethers catalyzed by ferric chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Buten-1-ol is employed in a study of the Mn-catalyzed hydrohydrazination of olefins. Also used in a study of the conversion of propargylic acetates to ethers catalyzed by ferric chloride.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

3-Buten-1-ol
0.841
32°C (89°F)
1.421
5 g
1633504
Soluble in water.
C=CCCO
72.107
72.11
627-27-0
113°C to 114°C
C4H8O
MFCD00002959
UN1987
3-buten-1-ol, allylcarbinol, 3-butenyl alcohol, 1-buten-4-ol, vinylethyl alcohol, 3-butene-1-ol, ch2=chch2ch2oh, homoallyl alcohol, unii-3db2krm1i9, 1-hydroxy-3-butene
ZSPTYLOMNJNZNG-UHFFFAOYSA-N
but-3-en-1-ol
69389
≥98%

Chemical Identifiers

627-27-0
72.107
ZSPTYLOMNJNZNG-UHFFFAOYSA-N
69389
C=CCCO
C4H8O
MFCD00002959
3-buten-1-ol, allylcarbinol, 3-butenyl alcohol, 1-buten-4-ol, vinylethyl alcohol, 3-butene-1-ol, ch2=chch2ch2oh, homoallyl alcohol, unii-3db2krm1i9, 1-hydroxy-3-butene
but-3-en-1-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

EINECSNumber : 210-991-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only