missing translation for 'onlineSavingsMsg'
Learn More

3-Buten-1-ol, 98+%

Catalog No. AAL0076714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0076714 25 g
AAL0076706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0076714 Supplier Thermo Scientific Chemicals Supplier No. L0076714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 627-27-0 | C4H8O | 72.107 g/mol

3-Buten-1-ol is employed in a study of the Mn-catalyzed hydrohydrazination of olefins. Also used in a study of the conversion of propargylic acetates to ethers catalyzed by ferric chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Buten-1-ol is employed in a study of the Mn-catalyzed hydrohydrazination of olefins. Also used in a study of the conversion of propargylic acetates to ethers catalyzed by ferric chloride.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 627-27-0
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.107
MDL Number MFCD00002959
InChI Key ZSPTYLOMNJNZNG-UHFFFAOYSA-N
Synonym 3-buten-1-ol, allylcarbinol, 3-butenyl alcohol, 1-buten-4-ol, vinylethyl alcohol, 3-butene-1-ol, ch2=chch2ch2oh, homoallyl alcohol, unii-3db2krm1i9, 1-hydroxy-3-butene
PubChem CID 69389
IUPAC Name but-3-en-1-ol
SMILES C=CCCO

Specifications

Density 0.841
Boiling Point 113°C to 114°C
Flash Point 32°C (89°F)
Refractive Index 1.421
Quantity 25 g
UN Number UN1987
Beilstein 1633504
Solubility Information Soluble in water.
Formula Weight 72.11
Percent Purity ≥98%
Chemical Name or Material 3-Buten-1-ol
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 210-991-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only