missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromothioanisole 98.0+%, TCI America™
$178.95 - $521.94
Chemical Identifiers
| CAS | 33733-73-2 |
|---|---|
| Molecular Formula | C7H7BrS |
| Molecular Weight (g/mol) | 203.097 |
| MDL Number | MFCD00041395 |
| InChI Key | NKYFJZAKUPSUSH-UHFFFAOYSA-N |
| Synonym | 3-bromothioanisole, 1-bromo-3-methylthio benzene, 3-bromophenyl methyl sulfide, 3-bromophenyl methyl sulfane, 1-bromo-3-methylsulfanyl benzene, benzene, 1-bromo-3-methylthio, m-bromo methylthio benzene, 3-bromophenyl methyl sulphide, 1-bromo-3-methylsulfanyl-benzene, 3-bromothioanisol |
| PubChem CID | 2735627 |
| IUPAC Name | 1-bromo-3-methylsulfanylbenzene |
| SMILES | CSC1=CC(=CC=C1)Br |
Chemical Identifiers
| 33733-73-2 | |
| 203.097 | |
| NKYFJZAKUPSUSH-UHFFFAOYSA-N | |
| 2735627 | |
| CSC1=CC(=CC=C1)Br |
| C7H7BrS | |
| MFCD00041395 | |
| 3-bromothioanisole, 1-bromo-3-methylthio benzene, 3-bromophenyl methyl sulfide, 3-bromophenyl methyl sulfane, 1-bromo-3-methylsulfanyl benzene, benzene, 1-bromo-3-methylthio, m-bromo methylthio benzene, 3-bromophenyl methyl sulphide, 1-bromo-3-methylsulfanyl-benzene, 3-bromothioanisol | |
| 1-bromo-3-methylsulfanylbenzene |
Specifications
| 33733-73-2 | |
| 250°C | |
| MFCD00041395 | |
| 3-bromothioanisole, 1-bromo-3-methylthio benzene, 3-bromophenyl methyl sulfide, 3-bromophenyl methyl sulfane, 1-bromo-3-methylsulfanyl benzene, benzene, 1-bromo-3-methylthio, m-bromo methylthio benzene, 3-bromophenyl methyl sulphide, 1-bromo-3-methylsulfanyl-benzene, 3-bromothioanisol | |
| CSC1=CC(=CC=C1)Br | |
| 203.097 | |
| 203.10 | |
| Liquid |
| Yellow | |
| C7H7BrS | |
| 5 g | |
| NKYFJZAKUPSUSH-UHFFFAOYSA-N | |
| 1-bromo-3-methylsulfanylbenzene | |
| 2735627 | |
| ≥98.0% (GC) | |
| 3-Bromothioanisole |
Safety and Handling
TSCA : No