missing translation for 'onlineSavingsMsg'
Learn More

3-Bromotetrahydro-4H-pyran-4-one, 90%, Thermo Scientific Chemicals

Catalog No. p-7057176
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66595MD 250 mg
AAH6659503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66595MD Supplier Thermo Scientific Chemicals Supplier No. H66595MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98021-79-5
Molecular Formula C5H7BrO2
Molecular Weight (g/mol) 179.013
MDL Number MFCD13193743
InChI Key ZGULSQDBOSMIBZ-UHFFFAOYSA-N
Synonym 3-bromodihydro-2h-pyran-4 3h-one, 3-bromotetrahydropyran-4-one, 4h-pyran-4-one, 3-bromotetrahydro, 3-bromotetrahydro-4h-pyran-4-one, 3-bromo-tetrahydropyran-4-one, acmc-20a5hh, 3-bromo-tetrahydro-pyran-4-one
PubChem CID 22224541
IUPAC Name 3-bromooxan-4-one
SMILES C1COCC(C1=O)Br

Specifications

Quantity 250 mg
Beilstein 109139
Formula Weight 179.01
Percent Purity 90%
Physical Form Solid
Chemical Name or Material 3-Bromotetrahydro-4H-pyran-4-one
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only