accessibility menu, dialog, popup

UserName

CAS RN 98021-79-5

IUPAC Name: 3-bromooxan-4-one
Synonyms: 3-bromooxan-4-one
Molecular Weight (g/mol): 179.01
Molecular Formula: C5H7BrO2
InChi Key: ZGULSQDBOSMIBZ-UHFFFAOYNA-N
SMILES: BrC1COCCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results