missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopyruvic Acid 98.0+%, TCI America™

$178.95 - $596.49

Chemical Identifiers

CAS 1113-59-3
Molecular Formula C3H3BrO3
Molecular Weight (g/mol) 166.958
MDL Number MFCD00002587
InChI Key PRRZDZJYSJLDBS-UHFFFAOYSA-N
Synonym 3-bromopyruvic acid, bromopyruvic acid, bromopyruvate, 3-bromopyruvate, 3-bromo-2-oxopropionic acid, bromopyruvicacid, pyruvic acid, bromo, propanoic acid, 3-bromo-2-oxo, unii-63jmv04grk, .beta.-bromopyruvic acid
PubChem CID 70684
IUPAC Name 3-bromo-2-oxopropanoic acid
SMILES C(C(=O)C(=O)O)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B11535G
View Documents
TCI America
B11535G
5 g
Each for $178.95
Only null left
 
B115325G
View Documents
TCI America
B115325G
25 g
Each for $596.49
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1113-59-3
166.958
PRRZDZJYSJLDBS-UHFFFAOYSA-N
70684
C(C(=O)C(=O)O)Br
C3H3BrO3
MFCD00002587
3-bromopyruvic acid, bromopyruvic acid, bromopyruvate, 3-bromopyruvate, 3-bromo-2-oxopropionic acid, bromopyruvicacid, pyruvic acid, bromo, propanoic acid, 3-bromo-2-oxo, unii-63jmv04grk, .beta.-bromopyruvic acid
3-bromo-2-oxopropanoic acid

Specifications

1113-59-3
White-Yellow
MFCD00002587
3261
PRRZDZJYSJLDBS-UHFFFAOYSA-N
3-bromo-2-oxopropanoic acid
70684
≥98.0% (T)
3-Bromopyruvic Acid
77°C
C3H3BrO3
5 g
3-bromopyruvic acid, bromopyruvic acid, bromopyruvate, 3-bromopyruvate, 3-bromo-2-oxopropionic acid, bromopyruvicacid, pyruvic acid, bromo, propanoic acid, 3-bromo-2-oxo, unii-63jmv04grk, .beta.-bromopyruvic acid
C(C(=O)C(=O)O)Br
166.958
166.96
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : UZ0840000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'