Learn More
3-Bromopyridine, 98+%
CAS: 626-55-1 | C5H4BrN | 158.00 g/mol
Supplier: Thermo Scientific Chemicals A1289414
Description
3-Bromopyridine undergoes lithiation with LDA at low temperatures in the 4-position: Heterocycles, involved in formation of the Grignard by Mg exchange with i-PrMgCl and in transmetallation of the lithiated derivative with zinc chloride followed by treatment with an electrophile as a route to 4-substituted 3-bromopyridines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Bromopyridine undergoes lithiation with LDA at low temperatures in the 4-position: Heterocycles, involved in formation of the Grignard by Mg exchange with i-PrMgCl and in transmetallation of the lithiated derivative with zinc chloride followed by treatment with an electrophile as a route to 4-substituted 3-bromopyridines.
Solubility
Soluble in water (31 g/L at 20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong acids.
Specifications
| 3-Bromopyridine | |
| -27°C | |
| 172°C to 174°C | |
| Characteristic | |
| 1.57 | |
| 25 g | |
| 105880 | |
| Soluble in water (31g/L at 20°C). | |
| BrC1=CC=CN=C1 | |
| 158.00 | |
| CHEBI:51575 | |
| ≥98% |
| 626-55-1 | |
| 1.617 | |
| 72°C (181°F) | |
| C5H4BrN | |
| MFCD00006373 | |
| UN2810 | |
| 3-pyridyl bromide, pyridine, 3-bromo, 3-bromo pyridine, 3-bromo-pyridine, 5-bromopyridine, unii-xmn8h2xe9h, 5-bromo pyridine, xmn8h2xe9h, 3-pyridylbromide, 3-bromopryidine | |
| NYPYPOZNGOXYSU-UHFFFAOYSA-N | |
| 3-bromopyridine | |
| 12286 | |
| 158 |
Chemical Identifiers
| 626-55-1 | |
| 158.00 | |
| NYPYPOZNGOXYSU-UHFFFAOYSA-N | |
| 12286 | |
| 3-bromopyridine |
| C5H4BrN | |
| MFCD00006373 | |
| 3-pyridyl bromide, pyridine, 3-bromo, 3-bromo pyridine, 3-bromo-pyridine, 5-bromopyridine, unii-xmn8h2xe9h, 5-bromo pyridine, xmn8h2xe9h, 3-pyridylbromide, 3-bromopryidine | |
| CHEBI:51575 | |
| BrC1=CC=CN=C1 |
Safety and Handling
GHS H Statement
H311-H227-H302-H332-H315-H319-H335
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H301+H311+H331-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 210-952-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only