missing translation for 'onlineSavingsMsg'
Learn More

(3-Bromopropoxy)(tert-butyl)dimethylsilane 98.0+%, TCI America™

$158.44 - $544.30

Chemical Identifiers

CAS 89031-84-5
Molecular Formula C9H21BrOSi
Molecular Weight (g/mol) 253.255
MDL Number MFCD00216589
InChI Key QGMROEZDWJTIDW-UHFFFAOYSA-N
Synonym 3-bromopropoxy-tert-butyldimethylsilane, 3-bromopropoxy tert-butyl dimethylsilane, 3-bromopropoxy-t-butyldimethylsilane, 3-bromopropoxy-tert-butyl-dimethyl-silane, silane, 3-bromopropoxy 1,1-dimethylethyl dimethyl, acmc-209unq, t-butyl 3-bromopropoxy dimethylsilane, bromopropoxy-tert-butyldimethyl silane
PubChem CID 607486
IUPAC Name 3-bromopropoxy-tert-butyl-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)OCCCBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B53465G
View Documents
TCI America
B53465G
5 g
Each for $158.44
Only null left
 
B534625G
View Documents
TCI America
B534625G
25 g
Each for $544.30
Only null left
 
Description

Description

Specifications

Chemical Identifiers

89031-84-5
253.255
QGMROEZDWJTIDW-UHFFFAOYSA-N
607486
CC(C)(C)[Si](C)(C)OCCCBr
C9H21BrOSi
MFCD00216589
3-bromopropoxy-tert-butyldimethylsilane, 3-bromopropoxy tert-butyl dimethylsilane, 3-bromopropoxy-t-butyldimethylsilane, 3-bromopropoxy-tert-butyl-dimethyl-silane, silane, 3-bromopropoxy 1,1-dimethylethyl dimethyl, acmc-209unq, t-butyl 3-bromopropoxy dimethylsilane, bromopropoxy-tert-butyldimethyl silane
3-bromopropoxy-tert-butyl-dimethylsilane

Specifications

89031-84-5
90°C
MFCD00216589
3-bromopropoxy-tert-butyldimethylsilane, 3-bromopropoxy tert-butyl dimethylsilane, 3-bromopropoxy-t-butyldimethylsilane, 3-bromopropoxy-tert-butyl-dimethyl-silane, silane, 3-bromopropoxy 1,1-dimethylethyl dimethyl, acmc-209unq, t-butyl 3-bromopropoxy dimethylsilane, bromopropoxy-tert-butyldimethyl silane
CC(C)(C)[Si](C)(C)OCCCBr
253.255
253.26
Liquid
Yellow
C9H21BrOSi
5 g
QGMROEZDWJTIDW-UHFFFAOYSA-N
3-bromopropoxy-tert-butyl-dimethylsilane
607486
≥98.0% (GC)
(3-Bromopropoxy)(tert-butyl)dimethylsilane
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'