missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3-Bromopropoxy)(tert-butyl)dimethylsilane 98.0+%, TCI America™
Supplier: TCI America B534625G
Specifications
| (3-Bromopropoxy)(tert-butyl)dimethylsilane | |
| Yellow | |
| C9H21BrOSi | |
| 25 g | |
| QGMROEZDWJTIDW-UHFFFAOYSA-N | |
| 3-bromopropoxy-tert-butyl-dimethylsilane | |
| 607486 | |
| ≥98.0% (GC) |
| 89031-84-5 | |
| 90°C | |
| MFCD00216589 | |
| 3-bromopropoxy-tert-butyldimethylsilane, 3-bromopropoxy tert-butyl dimethylsilane, 3-bromopropoxy-t-butyldimethylsilane, 3-bromopropoxy-tert-butyl-dimethyl-silane, silane, 3-bromopropoxy 1,1-dimethylethyl dimethyl, acmc-209unq, t-butyl 3-bromopropoxy dimethylsilane, bromopropoxy-tert-butyldimethyl silane | |
| CC(C)(C)[Si](C)(C)OCCCBr | |
| 253.255 | |
| 253.26 | |
| Liquid |
Chemical Identifiers
| 89031-84-5 | |
| 253.255 | |
| QGMROEZDWJTIDW-UHFFFAOYSA-N | |
| 607486 | |
| CC(C)(C)[Si](C)(C)OCCCBr |
| C9H21BrOSi | |
| MFCD00216589 | |
| 3-bromopropoxy-tert-butyldimethylsilane, 3-bromopropoxy tert-butyl dimethylsilane, 3-bromopropoxy-t-butyldimethylsilane, 3-bromopropoxy-tert-butyl-dimethyl-silane, silane, 3-bromopropoxy 1,1-dimethylethyl dimethyl, acmc-209unq, t-butyl 3-bromopropoxy dimethylsilane, bromopropoxy-tert-butyldimethyl silane | |
| 3-bromopropoxy-tert-butyl-dimethylsilane |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator