missing translation for 'onlineSavingsMsg'
Learn More

3-Bromophenylhydrazine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7123200
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
B38475G 5 g
B384725G 25 g
2 options

Catalog No. B38475G

Supplier: TCI America B38475G

Only null left

Chemical Identifiers

CAS 27246-81-7
Molecular Formula C6H8BrClN2
Molecular Weight (g/mol) 223.50
MDL Number MFCD00012933
InChI Key RPYIPFXHIKXRKS-UHFFFAOYSA-N
Synonym 3-bromophenylhydrazine hydrochloride, 3-bromophenyl hydrazine hydrochloride, 1-3-bromophenyl hydrazine hydrochloride, m-bromophenyl hydrazine hydrochloride, hydrazine, m-bromophenyl-, monohydrochloride, 3-bromophenylhydrazinehydrochloride, 3-bromophenyl hydrazine hcl, hydrazine, 3-bromophenyl-, monohydrochloride, pubchem23212, acmc-1clhx
PubChem CID 33757
IUPAC Name hydrogen (3-bromophenyl)hydrazine chloride
SMILES [H+].[Cl-].NNC1=CC=CC(Br)=C1

Specifications

Melting Point 225°C
Quantity 5 g
Formula Weight 223.50
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromophenylhydrazine Hydrochloride
RTECSNumber MV0815000
TSCA No