missing translation for 'onlineSavingsMsg'
Learn More

3-Bromophenylhydrazine hydrochloride, 98%

Catalog No. AC167350050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167350050 5 g Glass bottle
AC167350250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC167350050 Supplier Thermo Scientific Chemicals Supplier No. 167350050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 27246-81-7 | C6H8BrClN2 | 223.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27246-81-7
Molecular Formula C6H8BrClN2
Molecular Weight (g/mol) 223.50
MDL Number MFCD00012933
InChI Key RPYIPFXHIKXRKS-UHFFFAOYSA-N
Synonym 3-bromophenylhydrazine hydrochloride, 3-bromophenyl hydrazine hydrochloride, 1-3-bromophenyl hydrazine hydrochloride, m-bromophenyl hydrazine hydrochloride, hydrazine, m-bromophenyl-, monohydrochloride, 3-bromophenylhydrazinehydrochloride, 3-bromophenyl hydrazine hcl, hydrazine, 3-bromophenyl-, monohydrochloride, pubchem23212, acmc-1clhx
PubChem CID 33757
IUPAC Name (3-bromophenyl)hydrazine;hydrochloride
SMILES [H+].[Cl-].NNC1=CC=CC(Br)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 225°C to 231°C (decomposition)
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula BrC6H4NHNH2·HCl
Quantity 5 g
Beilstein 15, 433
Formula Weight 223.51
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material 3-Bromophenylhydrazine hydrochloride
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only