missing translation for 'onlineSavingsMsg'
Learn More

3-Bromophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. B28901G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B28901G Supplier TCI America Supplier No. B28901G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 89598-96-9
Molecular Formula C6H6BBrO2
Molecular Weight (g/mol) 200.83
MDL Number MFCD00239386
InChI Key AFSSVCNPDKKSRR-UHFFFAOYSA-N
Synonym 3-bromophenyl boronic acid, 3-bromobenzeneboronic acid, 3-bromophenylboronicacid, boronic acid, 3-bromophenyl, pubchem1758, 3-bromo-phenylboronic acid, acmc-209r1w, 3-bromo-benzeneboronic acid, 3-bromobenzene boronic acid
PubChem CID 2734318
IUPAC Name (3-bromophenyl)boronic acid
SMILES OB(O)C1=CC=CC(Br)=C1

Specifications

Melting Point 168°C
Color White-Yellow
Quantity 1 g
Formula Weight 200.83
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromophenylboronic Acid (contains varying amounts of Anhydride)