missing translation for 'onlineSavingsMsg'
Learn More

3-Bromophenylacetonitrile, 97%

Catalog No. p-7024924
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2018906 5 g
AAB2018914 25 g
AAB2018922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2018906 Supplier Thermo Scientific Chemicals Supplier No. B2018906

CAS: 31938-07-5 | C8H6BrN | 196.047 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31938-07-5
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.047
MDL Number MFCD00001906
InChI Key UUZYFBXKWIQKTF-UHFFFAOYSA-N
Synonym 3-bromophenylacetonitrile, 3-bromobenzyl cyanide, 2-3-bromophenyl acetonitrile, m-bromobenzyl cyanide, benzeneacetonitrile, 3-bromo, 3-bromobenzylcyanide, 3-bromobenzeneacetonitrile, 3-bromophenyl acetonitrile, m-bromophenylacetonitrile, 2-3-bromophenyl ethanenitrile
PubChem CID 36023
IUPAC Name 2-(3-bromophenyl)acetonitrile
SMILES C1=CC(=CC(=C1)Br)CC#N

Specifications

Melting Point 25°C to 28°C
Boiling Point 275°C
Flash Point 126°C (259°F)
Refractive Index 1.57
Quantity 5 g
UN Number UN3449
Beilstein 2355439
Formula Weight 196.05
Percent Purity 97%
Chemical Name or Material 3-Bromophenylacetonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H300-H310-H330-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: BROMOBENZYL CYANIDES, SOLID
EINECSNumber 250-867-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only