missing translation for 'onlineSavingsMsg'
Learn More

3-Bromophenylacetic acid, 98+%

Catalog No. AAB2075806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2075806 5 g
AAB2075814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2075806 Supplier Thermo Scientific Chemicals Supplier No. B2075806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1878-67-7 | C8H7BrO2 | 215.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1878-67-7
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00004330
InChI Key KYNNBXCGXUOREX-UHFFFAOYSA-N
Synonym 3-bromophenylacetic acid, 2-3-bromophenyl acetic acid, 3-bromophenyl acetic acid, m-bromophenylacetic acid, benzeneacetic acid, 3-bromo, 3-bromophenylaceticacid, 3-bromo-phenylacetic acid, 3-bromphenylacetic acid, pubchem4244
PubChem CID 74653
IUPAC Name 2-(3-bromophenyl)acetic acid
SMILES OC(=O)CC1=CC=CC(Br)=C1

Specifications

Melting Point 100°C to 102°C
Quantity 5 g
Beilstein 2045104
Formula Weight 215.05
Percent Purity ≥98%
Chemical Name or Material 3-Bromophenylacetic acid
EINECSNumber 217-522-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only