missing translation for 'onlineSavingsMsg'
Learn More

3-Bromophenyl Methyl Sulfone 98.0+%, TCI America™

Catalog No. B38935G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38935G 5 g
B389325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38935G Supplier TCI America Supplier No. B38935G
Only null left

Chemical Identifiers

CAS 34896-80-5
Molecular Formula C7H7BrO2S
Molecular Weight (g/mol) 235.10
MDL Number MFCD03095326
InChI Key WBOMXUMQOVQNKT-UHFFFAOYSA-N
PubChem CID 2757146
IUPAC Name 1-bromo-3-methanesulfonylbenzene
SMILES CS(=O)(=O)C1=CC=CC(Br)=C1

Specifications

Melting Point 105°C
Color White-Yellow
Quantity 5 g
Formula Weight 235.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromophenyl Methyl Sulfone