missing translation for 'onlineSavingsMsg'
Learn More

(3-Bromophenyl)diphenylphosphine Oxide 98.0+%, TCI America™

Catalog No. p-7124110
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5530200MG 200 mg
B55301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5530200MG Supplier TCI America Supplier No. B5530200MG
Only null left

Chemical Identifiers

CAS 10212-04-1
Molecular Formula C18H14BrOP
Molecular Weight (g/mol) 357.187
InChI Key GZZLAPUQZCXKKT-UHFFFAOYSA-N
PubChem CID 85529707
IUPAC Name 1-bromo-3-diphenylphosphorylbenzene
SMILES C1=CC=C(C=C1)P(=O)(C2=CC=CC=C2)C3=CC(=CC=C3)Br

Specifications

Melting Point 95°C
Color White
Quantity 200 mg
Formula Weight 357.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (3-Bromophenyl)diphenylphosphine Oxide