accessibility menu, dialog, popup

UserName

POBr

CAS RN 10212-04-1

IUPAC Name: 1-bromo-3-(diphenylphosphoroso)benzene
Synonyms: 1-bromo-3-(diphenylphosphoroso)benzene
Molecular Weight (g/mol): 357.19
Molecular Formula: C18H14BrOP
InChi Key: GZZLAPUQZCXKKT-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)P(=O)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results