missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromophenetole 98.0+%, TCI America™
$59.65 - $165.90
Chemical Identifiers
| CAS | 2655-84-7 |
|---|---|
| Molecular Formula | C8H9BrO |
| Molecular Weight (g/mol) | 201.06 |
| MDL Number | MFCD00274287 |
| InChI Key | LQBMPJSTUHWGDE-UHFFFAOYSA-N |
| Synonym | 3-bromophenetole, benzene, 1-bromo-3-ethoxy, m-bromophenetole, 3-bromophenyl ethyl ether, 1-bromo-3-ethoxy-benzene, 3-bromoethoxybenzene, 3-bromphenyl ethyl ether, phenetole, m-bromo, m-bromphenetol, 3-ethoxybromobenzene |
| PubChem CID | 75867 |
| IUPAC Name | 1-bromo-3-ethoxybenzene |
| SMILES | CCOC1=CC=CC(Br)=C1 |
Chemical Identifiers
| 2655-84-7 | |
| 201.06 | |
| LQBMPJSTUHWGDE-UHFFFAOYSA-N | |
| 75867 | |
| CCOC1=CC=CC(Br)=C1 |
| C8H9BrO | |
| MFCD00274287 | |
| 3-bromophenetole, benzene, 1-bromo-3-ethoxy, m-bromophenetole, 3-bromophenyl ethyl ether, 1-bromo-3-ethoxy-benzene, 3-bromoethoxybenzene, 3-bromphenyl ethyl ether, phenetole, m-bromo, m-bromphenetol, 3-ethoxybromobenzene | |
| 1-bromo-3-ethoxybenzene |
Specifications
| 2655-84-7 | |
| 231°C | |
| MFCD00274287 | |
| 3-bromophenetole, benzene, 1-bromo-3-ethoxy, m-bromophenetole, 3-bromophenyl ethyl ether, 1-bromo-3-ethoxy-benzene, 3-bromoethoxybenzene, 3-bromphenyl ethyl ether, phenetole, m-bromo, m-bromphenetol, 3-ethoxybromobenzene | |
| CCOC1=CC=CC(Br)=C1 | |
| 201.06 | |
| 201.06 | |
| Liquid |
| Yellow | |
| C8H9BrO | |
| 5 g | |
| LQBMPJSTUHWGDE-UHFFFAOYSA-N | |
| 1-bromo-3-ethoxybenzene | |
| 75867 | |
| ≥98.0% (GC) | |
| 3-Bromophenetole |
Safety and Handling
EINECSNumber : (3)-0845
TSCA : No