missing translation for 'onlineSavingsMsg'
Learn More

3-Bromophenetole, 98%

Catalog No. AAB2141809
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2141809 10 g
AAB2141818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2141809 Supplier Thermo Scientific Chemicals Supplier No. B2141809
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2655-84-7 | C8H9BrO | 201.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2655-84-7
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.06
MDL Number MFCD00274287
InChI Key LQBMPJSTUHWGDE-UHFFFAOYSA-N
Synonym 3-bromophenetole, benzene, 1-bromo-3-ethoxy, m-bromophenetole, 3-bromophenyl ethyl ether, 1-bromo-3-ethoxy-benzene, 3-bromoethoxybenzene, 3-bromphenyl ethyl ether, phenetole, m-bromo, m-bromphenetol, 3-ethoxybromobenzene
PubChem CID 75867
IUPAC Name 1-bromo-3-ethoxybenzene
SMILES CCOC1=CC=CC(Br)=C1

Specifications

Density 1.41
Boiling Point 231°C
Flash Point 83°C (182°F)
Refractive Index 1.548
Quantity 10 g
Formula Weight 201.06
Percent Purity 98%
Chemical Name or Material 3-Bromophenetole
EINECSNumber 220-186-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only