missing translation for 'onlineSavingsMsg'
Learn More

3-Bromophenanthrene 98.0+%, TCI America™

Catalog No. B2387100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2387100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B2387100MG Supplier TCI America Supplier No. B2387100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 715-50-4
Molecular Formula C14H9Br
Molecular Weight (g/mol) 257.13
MDL Number MFCD06797074
InChI Key BNGNNFQSUWVWCW-UHFFFAOYSA-N
PubChem CID 96767
IUPAC Name 3-bromophenanthrene
SMILES C1=CC=C2C(=C1)C=CC3=C2C=C(C=C3)Br

Specifications

Melting Point 83°C
Quantity 100 mg
Formula Weight 257.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromophenanthrene