accessibility menu, dialog, popup

UserName

Br

CAS RN 715-50-4

IUPAC Name: 3-bromophenanthrene
Synonyms: 3-bromophenanthrene
Molecular Weight (g/mol): 257.13
Molecular Formula: C14H9Br
InChi Key: BNGNNFQSUWVWCW-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C=CC3=CC=CC=C3C2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results