missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromopentane 90.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 118°C |
| Quantity | 25 g |
| UN Number | 1993 |
| Formula Weight | 151.05 |
| Percent Purity | ≥90.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Bromopentane |
Safety and Handling
| EINECSNumber | (2)-0067 |
| TSCA | Yes |