missing translation for 'onlineSavingsMsg'
Learn More

3-Bromooxetane, 95%

CAS: 39267-79-3 | C3H5BrO | 136.98 g/mol

Supplier:  Thermo Scientific Chemicals H57437ME

 View more versions of this product

Catalog No. AAH57437ME


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Bromooxetane
1.793
48°C (118°F)
MFCD08544402
UN1993
SZTIZZFKWQWSSP-UHFFFAOYSA-N
3-bromooxetane
16244493
95%
39267-79-3
131°C
C3H5BrO
500 mg
3-bromo-oxetane, 3-bromo oxetane, 3-bromanyloxetane, 3-bromooxetane, acmc-209j4f, 2-bromo-1,3-epoxypropane, 5-n,n-dimethylamino-2-hydroxybenzaldehyde
BrC1COC1
136.98
136.98

Chemical Identifiers

39267-79-3
136.98
SZTIZZFKWQWSSP-UHFFFAOYSA-N
16244493
BrC1COC1
C3H5BrO
MFCD08544402
3-bromo-oxetane, 3-bromo oxetane, 3-bromanyloxetane, 3-bromooxetane, acmc-209j4f, 2-bromo-1,3-epoxypropane, 5-n,n-dimethylamino-2-hydroxybenzaldehyde
3-bromooxetane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : No

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only