missing translation for 'onlineSavingsMsg'
Learn More

3-Bromooxetane, 95%

Catalog No. p-7038468
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH57437MD 250 mg
AAH57437ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH57437MD Supplier Thermo Scientific Chemicals Supplier No. H57437MD

CAS: 39267-79-3 | C3H5BrO | 136.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39267-79-3
Molecular Formula C3H5BrO
Molecular Weight (g/mol) 136.98
MDL Number MFCD08544402
InChI Key SZTIZZFKWQWSSP-UHFFFAOYSA-N
Synonym 3-bromo-oxetane, 3-bromo oxetane, 3-bromanyloxetane, 3-bromooxetane, acmc-209j4f, 2-bromo-1,3-epoxypropane, 5-n,n-dimethylamino-2-hydroxybenzaldehyde
PubChem CID 16244493
IUPAC Name 3-bromooxetane
SMILES BrC1COC1

Specifications

Density 1.793
Boiling Point 131°C
Flash Point 48°C (118°F)
Quantity 250 mg
UN Number UN1993
Formula Weight 136.98
Percent Purity 95%
Chemical Name or Material 3-Bromooxetane
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only