missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Bromomethyl)pyridine hydrobromide, 97%
CAS: 4916-55-6 | C6H6BrN·HBr | 252.93 g/mol
$435.07 - $435.07
Chemical Identifiers
| CAS | 4916-55-6 |
|---|---|
| Molecular Formula | C6H6BrN·HBr |
| Molecular Weight (g/mol) | 252.93 |
| MDL Number | MFCD01321314 |
| InChI Key | FNHPUOJKUXFUKN-UHFFFAOYSA-N |
| Synonym | 3-bromomethyl pyridine hydrobromide, 3-picolyl bromide hydrobromide, 3-bromomethylpyridine hydrobromide, 3-bromomethyl-pyridine hydrobromide, pyridine, 3-bromomethyl-, hydrobromide, pubchem17664, acmc-209kf1, 3-brommethyl pyridine-hbr, 3-bromomethyl pyridine, bromide, 3-bromomethyl pyridinehydrobromide |
| PubChem CID | 12707037 |
| IUPAC Name | 3-(bromomethyl)pyridine;hydrobromide |
| SMILES | C1=CC(=CN=C1)CBr.Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC397140050
|
Thermo Scientific Chemicals
397140050 |
5 g |
Each for $435.07
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4916-55-6 | |
| 252.93 | |
| FNHPUOJKUXFUKN-UHFFFAOYSA-N | |
| 12707037 | |
| C1=CC(=CN=C1)CBr.Br |
| C6H6BrN·HBr | |
| MFCD01321314 | |
| 3-bromomethyl pyridine hydrobromide, 3-picolyl bromide hydrobromide, 3-bromomethylpyridine hydrobromide, 3-bromomethyl-pyridine hydrobromide, pyridine, 3-bromomethyl-, hydrobromide, pubchem17664, acmc-209kf1, 3-brommethyl pyridine-hbr, 3-bromomethyl pyridine, bromide, 3-bromomethyl pyridinehydrobromide | |
| 3-(bromomethyl)pyridine;hydrobromide |
Specifications
| 4916-55-6 | |
| 97% | |
| C6H6BrN·HBr | |
| 5 g | |
| FNHPUOJKUXFUKN-UHFFFAOYSA-N | |
| 3-(bromomethyl)pyridine;hydrobromide | |
| 12707037 | |
| 97% |
| 150.0°C to 155.0°C | |
| Glass bottle | |
| MFCD01321314 | |
| 3-bromomethyl pyridine hydrobromide, 3-picolyl bromide hydrobromide, 3-bromomethylpyridine hydrobromide, 3-bromomethyl-pyridine hydrobromide, pyridine, 3-bromomethyl-, hydrobromide, pubchem17664, acmc-209kf1, 3-brommethyl pyridine-hbr, 3-bromomethyl pyridine, bromide, 3-bromomethyl pyridinehydrobromide | |
| C1=CC(=CN=C1)CBr.Br | |
| 252.93 | |
| 252.93 | |
| 3-(Bromomethyl)pyridine hydrobromide |
Safety and Handling
GHS Signal Word: Danger
RUO – Research Use Only