missing translation for 'onlineSavingsMsg'
Learn More

3-(Bromomethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. B38141G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B38141G Supplier TCI America Supplier No. B38141G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 3-(Bromomethyl)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 51323-43-4
Melting Point 216°C
Color White-Yellow
Molecular Formula C7H8BBrO2
Quantity 1 g
MDL Number MFCD01632207
UN Number 3261
Synonym 3-bromomethylphenylboronic acid, 3-bromomethyl phenylboronic acid, 3-bromomethyl benzeneboronic acid, 3-bromomethyl phenyl boronic acid, 3-bromomethylphenyl boronic acid, m-bromomethyl phenylboronic acid, boronic acid, 3-bromomethyl phenyl, pubchem7782, acmc-1aoze
InChI Key ATRFDLFMCLYROQ-UHFFFAOYSA-N
SMILES OB(O)C1=CC=CC(CBr)=C1
IUPAC Name [3-(bromomethyl)phenyl]boronic acid
Molecular Weight (g/mol) 214.85
PubChem CID 2773281
Formula Weight 214.85
Physical Form Crystalline Powder
Show More Show Less