Learn More
3-(Bromomethyl)phenylboronic acid, 97%
CAS: 51323-43-4 | C7H8BBrO2 | 214.85 g/mol
$84.10 - $84.10
Chemical Identifiers
| CAS | 51323-43-4 |
|---|---|
| Molecular Formula | C7H8BBrO2 |
| Molecular Weight (g/mol) | 214.85 |
| MDL Number | MFCD01632207 |
| InChI Key | ATRFDLFMCLYROQ-UHFFFAOYSA-N |
| Synonym | 3-bromomethylphenylboronic acid, 3-bromomethyl phenylboronic acid, 3-bromomethyl benzeneboronic acid, 3-bromomethyl phenyl boronic acid, 3-bromomethylphenyl boronic acid, m-bromomethyl phenylboronic acid, boronic acid, 3-bromomethyl phenyl, pubchem7782, acmc-1aoze |
| PubChem CID | 2773281 |
| IUPAC Name | [3-(bromomethyl)phenyl]boronic acid |
| SMILES | OB(O)C1=CC=CC(CBr)=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC430600010
|
Thermo Scientific Chemicals
430600010 |
1 g | Glass bottle |
Each for $84.10
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 51323-43-4 | |
| 214.85 | |
| ATRFDLFMCLYROQ-UHFFFAOYSA-N | |
| 2773281 | |
| OB(O)C1=CC=CC(CBr)=C1 |
| C7H8BBrO2 | |
| MFCD01632207 | |
| 3-bromomethylphenylboronic acid, 3-bromomethyl phenylboronic acid, 3-bromomethyl benzeneboronic acid, 3-bromomethyl phenyl boronic acid, 3-bromomethylphenyl boronic acid, m-bromomethyl phenylboronic acid, boronic acid, 3-bromomethyl phenyl, pubchem7782, acmc-1aoze | |
| [3-(bromomethyl)phenyl]boronic acid |
Specifications
| 51323-43-4 | |
| 100.0 | |
| Tan to White | |
| 96% min. (HPLC) | |
| C7H8BBrO2 | |
| 1 g | |
| ATRFDLFMCLYROQ-UHFFFAOYSA-N | |
| [3-(bromomethyl)phenyl]boronic acid | |
| 2773281 | |
| 97% | |
| 3-(Bromomethyl)phenylboronic acid |
| 96.0 | |
| 209.0°C to 211.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD01632207 | |
| 3-bromomethylphenylboronic acid, 3-bromomethyl phenylboronic acid, 3-bromomethyl benzeneboronic acid, 3-bromomethyl phenyl boronic acid, 3-bromomethylphenyl boronic acid, m-bromomethyl phenylboronic acid, boronic acid, 3-bromomethyl phenyl, pubchem7782, acmc-1aoze | |
| OB(O)C1=CC=CC(CBr)=C1 | |
| 214.85 | |
| 214.85 | |
| Crystalline Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO – Research Use Only