missing translation for 'onlineSavingsMsg'
Learn More

3-(Bromomethyl)phenoxyacetic acid, 97%

Catalog No. AAL1802903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1802903 1 g
AAL1802906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1802903 Supplier Thermo Scientific Chemicals Supplier No. L1802903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 136645-25-5 | C9H9BrO3 | 245.072 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 136645-25-5
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.072
MDL Number MFCD02093985
InChI Key MSTODKGDFXWAIO-UHFFFAOYSA-N
Synonym 3-bromomethyl phenoxyacetic acid, 2-3-bromomethyl phenoxy acetic acid, acmc-1c8fy
PubChem CID 7010327
IUPAC Name 2-[3-(bromomethyl)phenoxy]acetic acid
SMILES C1=CC(=CC(=C1)OCC(=O)O)CBr

Specifications

Melting Point 124°C to 128°C
Quantity 1 g
UN Number UN3261
Formula Weight 245.07
Percent Purity 97%
Chemical Name or Material 3-(Bromomethyl)phenoxyacetic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only