Learn More
3-(Bromomethyl)benzonitrile, 98%
CAS: 28188-41-2 | C8H6BrN | 196.05 g/mol
Supplier: Thermo Scientific Chemicals A1524414
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3-(Bromomethyl)benzonitrile | |
| 92°C to 96°C | |
| MFCD00001809 | |
| UN2923 | |
| 3-bromomethyl benzonitrile, 3-cyanobenzyl bromide, alpha-bromo-m-tolunitrile, m-cyanobenzyl bromide, benzonitrile, 3-bromomethyl, m-bromomethyl benzonitrile, alpha-bromo-m-toluonitrile, m-tolunitrile, .alpha.-bromo, a-bromo-m-tolunitrile, 3-bromomethyl-benzonitrile | |
| BrCC1=CC=CC(=C1)C#N | |
| 196.05 | |
| 196.05 |
| 28188-41-2 | |
| C8H6BrN | |
| 25 g | |
| 742356 | |
| CVKOOKPNCVYHNY-UHFFFAOYSA-N | |
| 3-(bromomethyl)benzonitrile | |
| 97249 | |
| 98% |
Chemical Identifiers
| 28188-41-2 | |
| 196.05 | |
| CVKOOKPNCVYHNY-UHFFFAOYSA-N | |
| 97249 | |
| BrCC1=CC=CC(=C1)C#N |
| C8H6BrN | |
| MFCD00001809 | |
| 3-bromomethyl benzonitrile, 3-cyanobenzyl bromide, alpha-bromo-m-tolunitrile, m-cyanobenzyl bromide, benzonitrile, 3-bromomethyl, m-bromomethyl benzonitrile, alpha-bromo-m-toluonitrile, m-tolunitrile, .alpha.-bromo, a-bromo-m-tolunitrile, 3-bromomethyl-benzonitrile | |
| 3-(bromomethyl)benzonitrile |
Safety and Handling
GHS H Statement
H301-H311-H331-H314-H318-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber : 248-890-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only