missing translation for 'onlineSavingsMsg'
Learn More

3-(Bromomethyl)benzonitrile, 98%

CAS: 28188-41-2 | C8H6BrN | 196.05 g/mol

Supplier:  Thermo Scientific Chemicals A1524406

 View more versions of this product

Catalog No. AAA1524406


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-(Bromomethyl)benzonitrile
92°C to 96°C
MFCD00001809
UN2923
3-bromomethyl benzonitrile, 3-cyanobenzyl bromide, alpha-bromo-m-tolunitrile, m-cyanobenzyl bromide, benzonitrile, 3-bromomethyl, m-bromomethyl benzonitrile, alpha-bromo-m-toluonitrile, m-tolunitrile, .alpha.-bromo, a-bromo-m-tolunitrile, 3-bromomethyl-benzonitrile
BrCC1=CC=CC(=C1)C#N
196.05
196.05
28188-41-2
C8H6BrN
5 g
742356
CVKOOKPNCVYHNY-UHFFFAOYSA-N
3-(bromomethyl)benzonitrile
97249
98%

Chemical Identifiers

28188-41-2
196.05
CVKOOKPNCVYHNY-UHFFFAOYSA-N
97249
BrCC1=CC=CC(=C1)C#N
C8H6BrN
MFCD00001809
3-bromomethyl benzonitrile, 3-cyanobenzyl bromide, alpha-bromo-m-tolunitrile, m-cyanobenzyl bromide, benzonitrile, 3-bromomethyl, m-bromomethyl benzonitrile, alpha-bromo-m-toluonitrile, m-tolunitrile, .alpha.-bromo, a-bromo-m-tolunitrile, 3-bromomethyl-benzonitrile
3-(bromomethyl)benzonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H314-H318-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

EINECSNumber : 248-890-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only