missing translation for 'onlineSavingsMsg'
Learn More

3-(Bromomethyl)benzonitrile, 98%

Catalog No. AAA1524422
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1524422 100 g
AAA1524406 5 g
AAA1524414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1524422 Supplier Thermo Scientific Chemicals Supplier No. A1524422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 28188-41-2 | C8H6BrN | 196.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 28188-41-2
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.05
MDL Number MFCD00001809
InChI Key CVKOOKPNCVYHNY-UHFFFAOYSA-N
Synonym 3-bromomethyl benzonitrile, 3-cyanobenzyl bromide, alpha-bromo-m-tolunitrile, m-cyanobenzyl bromide, benzonitrile, 3-bromomethyl, m-bromomethyl benzonitrile, alpha-bromo-m-toluonitrile, m-tolunitrile, .alpha.-bromo, a-bromo-m-tolunitrile, 3-bromomethyl-benzonitrile
PubChem CID 97249
IUPAC Name 3-(bromomethyl)benzonitrile
SMILES BrCC1=CC=CC(=C1)C#N

Specifications

Melting Point 92°C to 96°C
Quantity 100 g
UN Number UN2923
Beilstein 742356
Formula Weight 196.05
Percent Purity 98%
Chemical Name or Material 3-(Bromomethyl)benzonitrile
Hazard Category H314
Hazard Statement GHS H Statement
H301-H311-H331-H314-H318-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 248-890-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only