missing translation for 'onlineSavingsMsg'
Learn More

3-(Bromomethyl)-3-oxetanemethanol 97.0+%, TCI America™

Catalog No. B54421G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B54421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B54421G Supplier TCI America Supplier No. B54421G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22633-44-9
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD09800598
InChI Key SESXZSLSTRITGO-UHFFFAOYSA-N
Synonym 3-bromomethyl oxetan-3-yl methanol, 3-bromomethyl-3-oxetanemethanol, 3-bromomethyl-3-hydroxymethyloxetane, 3-oxetanemethanol, 3-bromomethyl, oxetane, 3-bromomethyl-3-hydroxymethyl, 3-bromomethyl-3-hydroxymethyl oxetane, 3-bromomethyl oxetan-3-yl methan-1-ol, 3-oxetanemethanol,3-bromomethyl, 2-bromomethyl-2-hydroxymethyl-1,3-epoxypropane
PubChem CID 529264
IUPAC Name [3-(bromomethyl)oxetan-3-yl]methanol
SMILES OCC1(CBr)COC1

Specifications

Color Yellow
Boiling Point 150°C
Quantity 1 g
Formula Weight 181.03
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Bromomethyl)-3-oxetanemethanol