accessibility menu, dialog, popup

UserName

CAS RN 22633-44-9

IUPAC Name: [3-(bromomethyl)oxetan-3-yl]methanol
Synonyms: [3-(bromomethyl)oxetan-3-yl]methanol
Molecular Weight (g/mol): 181.03
Molecular Formula: C5H9BrO2
InChi Key: SESXZSLSTRITGO-UHFFFAOYSA-N
SMILES: OCC1(CBr)COC1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)