missing translation for 'onlineSavingsMsg'
Learn More

3-Bromomethyl-3-oxetanemethanol, 95%

CAS: 22633-44-9 | C5H9BrO2 | 181.03 g/mol

Supplier:  Thermo Scientific Chemicals H57413ME

 View more versions of this product

Catalog No. AAH57413ME


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Bromomethyl-3-oxetanemethanol
1.598
100°C (212°F)
MFCD09800598
3-bromomethyl oxetan-3-yl methanol, 3-bromomethyl-3-oxetanemethanol, 3-bromomethyl-3-hydroxymethyloxetane, 3-oxetanemethanol, 3-bromomethyl, oxetane, 3-bromomethyl-3-hydroxymethyl, 3-bromomethyl-3-hydroxymethyl oxetane, 3-bromomethyl oxetan-3-yl methan-1-ol, 3-oxetanemethanol,3-bromomethyl, 2-bromomethyl-2-hydroxymethyl-1,3-epoxypropane
OCC1(CBr)COC1
181.03
181.03
22633-44-9
242°C
C5H9BrO2
500 mg
SESXZSLSTRITGO-UHFFFAOYSA-N
[3-(bromomethyl)oxetan-3-yl]methanol
529264
95%

Chemical Identifiers

22633-44-9
181.03
SESXZSLSTRITGO-UHFFFAOYSA-N
[3-(bromomethyl)oxetan-3-yl]methanol
C5H9BrO2
MFCD09800598
529264
OCC1(CBr)COC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only