missing translation for 'onlineSavingsMsg'
Learn More

3-Bromomethyl-3-oxetanemethanol, 95%

Catalog No. AAH57413MD
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH57413MD 250 mg
AAH57413ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH57413MD Supplier Thermo Scientific Chemicals Supplier No. H57413MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 22633-44-9 | C5H9BrO2 | 181.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22633-44-9
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD09800598
InChI Key SESXZSLSTRITGO-UHFFFAOYSA-N
PubChem CID 529264
IUPAC Name [3-(bromomethyl)oxetan-3-yl]methanol
SMILES OCC1(CBr)COC1

Specifications

Density 1.598
Boiling Point 242°C
Flash Point 100°C (212°F)
Quantity 250 mg
Synonym 3-bromomethyl oxetan-3-yl methanol, 3-bromomethyl-3-oxetanemethanol, 3-bromomethyl-3-hydroxymethyloxetane, 3-oxetanemethanol, 3-bromomethyl, oxetane, 3-bromomethyl-3-hydroxymethyl, 3-bromomethyl-3-hydroxymethyl oxetane, 3-bromomethyl oxetan-3-yl methan-1-ol, 3-oxetanemethanol,3-bromomethyl, 2-bromomethyl-2-hydroxymethyl-1,3-epoxypropane
Formula Weight 181.03
Percent Purity 95%
Chemical Name or Material 3-Bromomethyl-3-oxetanemethanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only