missing translation for 'onlineSavingsMsg'
Learn More

3-Bromomethyl-2(1H)-quinoxalinone, tech. 90%

Catalog No. AAH2716906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2716906 5 g
AAH2716914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2716906 Supplier Thermo Scientific Chemicals Supplier No. H2716906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 62235-61-4 | C9H7BrN2O | 239.072 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62235-61-4
Molecular Formula C9H7BrN2O
Molecular Weight (g/mol) 239.072
MDL Number MFCD02660613
InChI Key SOTAKCFOTFCVSS-UHFFFAOYSA-N
Synonym 3-bromomethyl quinoxalin-2-ol, 3-bromomethylquinoxalin-2-one, 3-bromomethyl-1h-quinoxalin-2-one, 3-bromomethyl-2-quinoxalinol, 3-bromomethyl-2 1h-quinoxalinone, 2 1h-quinoxalinone, 3-bromomethyl, 3-bromomethyl hydroquinoxalin-2-one, acmc-20agnv, 3-bromomethyl quinoxaline
PubChem CID 286254
IUPAC Name 3-(bromomethyl)-1H-quinoxalin-2-one
SMILES C1=CC=C2C(=C1)NC(=O)C(=N2)CBr

Specifications

Melting Point >300°C (decomposition)
Quantity 5 g
UN Number UN1759
Formula Weight 239.07
Percent Purity 90%
Grade Technical
Chemical Name or Material 3-Bromomethyl-2(1H)-quinoxalinone
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only