missing translation for 'onlineSavingsMsg'
Learn More

3-Bromoisobutyric Acid 97.0+%, TCI America™

Catalog No. B257125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B257125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B257125G Supplier TCI America Supplier No. B257125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56970-78-6
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.00
MDL Number MFCD01310530
InChI Key BUPXDXGYFXDDAA-UHFFFAOYNA-N
Synonym 3-Bromo-2-methylpropionic Acid
PubChem CID 304776
IUPAC Name 3-bromo-2-methylpropanoic acid
SMILES CC(CBr)C(O)=O

Specifications

Color Yellow
Boiling Point 231°C
Quantity 25 g
UN Number 3265
Formula Weight 167.00
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Bromoisobutyric Acid
EINECSNumber (2)-3494
TSCA No