missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromoimidazo[1,2-a]pyridine, 97%, Thermo Scientific™
$79.99 - $79.99
Chemical Identifiers
| CAS | 4926-47-0 |
|---|---|
| Molecular Formula | C7H5BrN2 |
| Molecular Weight (g/mol) | 197.035 |
| InChI Key | APYSHMNJHJRIDR-UHFFFAOYSA-N |
| PubChem CID | 327951 |
| IUPAC Name | 3-bromoimidazo[1,2-a]pyridine |
| SMILES | C1=CC2=NC=C(N2C=C1)Br |
Chemical Identifiers
| 4926-47-0 | |
| 197.035 | |
| 327951 | |
| C1=CC2=NC=C(N2C=C1)Br |
| C7H5BrN2 | |
| APYSHMNJHJRIDR-UHFFFAOYSA-N | |
| 3-bromoimidazo[1,2-a]pyridine |
Specifications
| 4926-47-0 | |
| Amber glass bottle | |
| 1 g | |
| APYSHMNJHJRIDR-UHFFFAOYSA-N | |
| 3-bromoimidazo[1,2-a]pyridine | |
| 327951 | |
| 97% |
| 97% | |
| C7H5BrN2 | |
| 3-bromoimidazo 1,2-a pyridine, 3-bromo-imidazo 1,2-a pyridine, 3-bromopyrimidazole, imidazo 1,2-a pyridine, 3-bromo, 3-bromoimidazolo 1,2-a pyridine, 3-bromoh-imidazo 1,2-a pyridine, pubchem17183, acmc-209kfj, buttpark 96\50-11, imidazo 1,2-a pyridine,3-bromo | |
| C1=CC2=NC=C(N2C=C1)Br | |
| 197.035 | |
| 197.03 | |
| 3-Bromoimidazo[1, 2-a]pyridine |