accessibility menu, dialog, popup

UserName

CAS RN 4926-47-0

IUPAC Name: 3-bromoimidazo[1,2-a]pyridine
Synonyms: 3-bromoimidazo[1,2-a]pyridine
Molecular Weight (g/mol): 197.04
Molecular Formula: C7H5BrN2
InChi Key: APYSHMNJHJRIDR-UHFFFAOYSA-N
SMILES: BrC1=CN=C2C=CC=CN12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results