missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromofluorobenzene 98.0+%, TCI America™
$77.00 - $396.59
Chemical Identifiers
| CAS | 1073-06-9 |
|---|---|
| Molecular Formula | C6H4BrF |
| Molecular Weight (g/mol) | 175 |
| MDL Number | MFCD00000326 |
| InChI Key | QDFKKJYEIFBEFC-UHFFFAOYSA-N |
| Synonym | 3-bromofluorobenzene, m-bromofluorobenzene, m-fluorobromobenzene, benzene, 1-bromo-3-fluoro, 3-bromfluorbenzen, 3-fluorobromobenzene, m-fluorophenyl bromide, 1-fluoro-3-bromobenzene, 3-fluoro bromobenzene, benzene, 1-fluoro-3-bromo |
| PubChem CID | 14082 |
| IUPAC Name | 1-bromo-3-fluorobenzene |
| SMILES | C1=CC(=CC(=C1)Br)F |
Chemical Identifiers
| 1073-06-9 | |
| 175 | |
| QDFKKJYEIFBEFC-UHFFFAOYSA-N | |
| 14082 | |
| C1=CC(=CC(=C1)Br)F |
| C6H4BrF | |
| MFCD00000326 | |
| 3-bromofluorobenzene, m-bromofluorobenzene, m-fluorobromobenzene, benzene, 1-bromo-3-fluoro, 3-bromfluorbenzen, 3-fluorobromobenzene, m-fluorophenyl bromide, 1-fluoro-3-bromobenzene, 3-fluoro bromobenzene, benzene, 1-fluoro-3-bromo | |
| 1-bromo-3-fluorobenzene |
Specifications
| 1073-06-9 | |
| 147°C | |
| MFCD00000326 | |
| 1993 | |
| QDFKKJYEIFBEFC-UHFFFAOYSA-N | |
| 1-bromo-3-fluorobenzene | |
| 14082 | |
| ≥98.0% (GC) | |
| 3-Bromofluorobenzene |
| Yellow | |
| C6H4BrF | |
| 25 g | |
| 3-bromofluorobenzene, m-bromofluorobenzene, m-fluorobromobenzene, benzene, 1-bromo-3-fluoro, 3-bromfluorbenzen, 3-fluorobromobenzene, m-fluorophenyl bromide, 1-fluoro-3-bromobenzene, 3-fluoro bromobenzene, benzene, 1-fluoro-3-bromo | |
| C1=CC(=CC(=C1)Br)F | |
| 175 | |
| 175.00 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-4286
RTECSNumber : DA1225000
TSCA : Yes