missing translation for 'onlineSavingsMsg'
Learn More

3-Bromofluorobenzene, 99%

Catalog No. p-74613
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC107000250 25 mL
AC107001000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC107000250 Supplier Thermo Scientific Chemicals Supplier No. 107000250

CAS: 1073-06-9 | C6H4BrF | 175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1073-06-9
Molecular Formula C6H4BrF
Molecular Weight (g/mol) 175
MDL Number MFCD00000326
InChI Key QDFKKJYEIFBEFC-UHFFFAOYSA-N
Synonym 3-bromofluorobenzene, m-bromofluorobenzene, m-fluorobromobenzene, benzene, 1-bromo-3-fluoro, 3-bromfluorbenzen, 3-fluorobromobenzene, m-fluorophenyl bromide, 1-fluoro-3-bromobenzene, 3-fluoro bromobenzene, benzene, 1-fluoro-3-bromo
PubChem CID 14082
IUPAC Name 1-bromo-3-fluorobenzene
SMILES C1=CC(=CC(=C1)Br)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.5670g/mL
Boiling Point 150°C
Flash Point 38°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Refractive Index 1.5260 to 1.528
Linear Formula BrC6H4F
Quantity 25 mL
Specific Gravity 1.567
Solubility Information Solubility in water: insoluble in water. Other solubilities: organic soluble/miscible with most solvents
Formula Weight 175
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Bromofluorobenzene, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-023-0
RTECSNumber DA1225000
TSCA TSCA

RUO – Research Use Only