missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromodibenzothiophene 5,5-Dioxide 98.0+%, TCI America™
$346.71 - $1032.68
Chemical Identifiers
| CAS | 116668-69-0 |
|---|---|
| Molecular Formula | C12H7BrO2S |
| Molecular Weight (g/mol) | 295.15 |
| InChI Key | XHRCPNQYOKMDIP-UHFFFAOYSA-N |
| Synonym | 3-Bromodibenzothiophene Sulfone, 3-Bromodibenzosulfolane |
| PubChem CID | 21818589 |
| IUPAC Name | 3-bromodibenzothiophene 5,5-dioxide |
| SMILES | C1=CC=C2C(=C1)C3=C(S2(=O)=O)C=C(C=C3)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
B5268200MG
|
TCI America
B5268200MG |
200 mg |
Each for $346.71
|
|
|||||
|
B52681G
|
TCI America
B52681G |
1 g |
Each for $1,032.68
|
|
|||||
Chemical Identifiers
| 116668-69-0 | |
| 295.15 | |
| 3-Bromodibenzothiophene Sulfone, 3-Bromodibenzosulfolane | |
| 3-bromodibenzothiophene 5,5-dioxide |
| C12H7BrO2S | |
| XHRCPNQYOKMDIP-UHFFFAOYSA-N | |
| 21818589 | |
| C1=CC=C2C(=C1)C3=C(S2(=O)=O)C=C(C=C3)Br |
Specifications
| 116668-69-0 | |
| C12H7BrO2S | |
| 3-Bromodibenzothiophene Sulfone, 3-Bromodibenzosulfolane | |
| C1=CC=C2C(=C1)C3=C(S2(=O)=O)C=C(C=C3)Br | |
| 295.15 | |
| 295.15 | |
| Crystalline Powder |
| 228°C | |
| 200 mg | |
| XHRCPNQYOKMDIP-UHFFFAOYSA-N | |
| 3-bromodibenzothiophene 5,5-dioxide | |
| 21818589 | |
| ≥98.0% (GC) | |
| 3-Bromodibenzothiophene 5,5-Dioxide |
Safety and Handling
TSCA : No