accessibility menu, dialog, popup

UserName

Enjoy Up to 50% Off Spotlight Deals All Year Long

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 116668-69-0

SOOBr

CAS RN 116668-69-0

IUPAC Name: 11-bromo-8λ⁶-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2,4,6,10,12-hexaene-8,8-dione
Synonyms: 11-bromo-8λ⁶-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2,4,6,10,12-hexaene-8,8-dione
Molecular Weight (g/mol): 295.15
Molecular Formula: C12H7BrO2S
InChi Key: XHRCPNQYOKMDIP-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(C=C1)C1=CC=CC=C1S2(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.