Learn More
3-Bromocarbazole, 98%
CAS: 1592-95-6 | C12H8BrN | 246.107 g/mol
$59.30 - $669.05
Chemical Identifiers
| CAS | 1592-95-6 |
|---|---|
| Molecular Formula | C12H8BrN |
| Molecular Weight (g/mol) | 246.107 |
| MDL Number | MFCD00222621 |
| InChI Key | LTBWKAYPXIIVPC-UHFFFAOYSA-N |
| Synonym | 3-bromocarbazole, 9h-carbazole, 3-bromo, 3-brominecarbazole, 3bromo-9h-carbazole, pubchem9950, maybridge1_004311, acmc-1bpt1, 3-bromocarbazole gc |
| PubChem CID | 252446 |
| IUPAC Name | 3-bromo-9H-carbazole |
| SMILES | C1=CC=C2C(=C1)C3=C(N2)C=CC(=C3)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6443703
|
Thermo Scientific Chemicals
H6443703 |
1 g |
Each for $59.30
|
|
|||||
|
AAH6443706
|
Thermo Scientific Chemicals
H6443706 |
5 g |
Each for $185.36
|
|
|||||
|
AAH6443714
|
Thermo Scientific Chemicals
H6443714 |
25 g |
Each for $669.05
|
|
|||||
Description
Used as a intermediate for organic light-emitting diode(OLED) and pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as a intermediate for organic light-emitting diode(OLED) and pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
| 1592-95-6 | |
| 246.107 | |
| LTBWKAYPXIIVPC-UHFFFAOYSA-N | |
| 252446 | |
| C1=CC=C2C(=C1)C3=C(N2)C=CC(=C3)Br |
| C12H8BrN | |
| MFCD00222621 | |
| 3-bromocarbazole, 9h-carbazole, 3-bromo, 3-brominecarbazole, 3bromo-9h-carbazole, pubchem9950, maybridge1_004311, acmc-1bpt1, 3-bromocarbazole gc | |
| 3-bromo-9H-carbazole |
Specifications
| 1592-95-6 | |
| White | |
| C12H8BrN | |
| 1 g | |
| 3-bromocarbazole, 9h-carbazole, 3-bromo, 3-brominecarbazole, 3bromo-9h-carbazole, pubchem9950, maybridge1_004311, acmc-1bpt1, 3-bromocarbazole gc | |
| LTBWKAYPXIIVPC-UHFFFAOYSA-N | |
| 3-bromo-9H-carbazole | |
| 252446 | |
| 98% | |
| 3-Bromocarbazole |
| 195°C to 199°C | |
| 160°C (6 mmHg) | |
| MFCD00222621 | |
| 142195 | |
| Slightly soluble in water. | |
| C1=CC=C2C(=C1)C3=C(N2)C=CC(=C3)Br | |
| 246.107 | |
| 246.11 | |
| Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only