missing translation for 'onlineSavingsMsg'
Learn More

3-Bromocarbazole, 98%

CAS: 1592-95-6 | C12H8BrN | 246.107 g/mol

Supplier:  Thermo Scientific Chemicals H6443706

 View more versions of this product

Catalog No. AAH6443706


Only null left
Explore available promotions

Description

Description

Used as a intermediate for organic light-emitting diode(OLED) and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a intermediate for organic light-emitting diode(OLED) and pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

3-Bromocarbazole
195°C to 199°C
160°C (6 mmHg)
MFCD00222621
142195
Slightly soluble in water.
C1=CC=C2C(=C1)C3=C(N2)C=CC(=C3)Br
246.107
246.11
Solid
1592-95-6
White
C12H8BrN
5 g
3-bromocarbazole, 9h-carbazole, 3-bromo, 3-brominecarbazole, 3bromo-9h-carbazole, pubchem9950, maybridge1_004311, acmc-1bpt1, 3-bromocarbazole gc
LTBWKAYPXIIVPC-UHFFFAOYSA-N
3-bromo-9H-carbazole
252446
98%

Chemical Identifiers

1592-95-6
246.107
LTBWKAYPXIIVPC-UHFFFAOYSA-N
252446
C1=CC=C2C(=C1)C3=C(N2)C=CC(=C3)Br
C12H8BrN
MFCD00222621
3-bromocarbazole, 9h-carbazole, 3-bromo, 3-brominecarbazole, 3bromo-9h-carbazole, pubchem9950, maybridge1_004311, acmc-1bpt1, 3-bromocarbazole gc
3-bromo-9H-carbazole
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only