missing translation for 'onlineSavingsMsg'
Learn More

(-)-3-Bromocamphor-8-sulfonic Acid Ammonium Salt 97.0+%, TCI America™

Catalog No. B44121G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44121G 1 g
B44125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44121G Supplier TCI America Supplier No. B44121G

Chemical Identifiers

CAS 55870-50-3
Molecular Formula C10H18BrNO4S
Molecular Weight (g/mol) 328.221
MDL Number MFCD00151070
InChI Key GFBVBBRNPGPROZ-ORUKMIDOSA-N
Synonym l--alpha-bromocamphor-pi-sulfonic acid, ammonium salt, ammonium--3-bromo-8-camphorsulfonate, 1s,3r,4r,7s-3-bromo-1,7-dimethyl-2-oxobicyclo 2.2.1 heptan-7-yl methanesulfonic acid amine, 1s---3-bromocamphor-8-sulfonic acid ammonium salt
PubChem CID 71306756
IUPAC Name azane;[(1R,2R,4S,7S)-2-bromo-4,7-dimethyl-3-oxo-7-bicyclo[2.2.1]heptanyl]methanesulfonic acid
SMILES CC12CCC(C1(C)CS(=O)(=O)O)C(C2=O)Br.N

Specifications

Melting Point 273°C
Quantity 1 g
Formula Weight 328.22
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (-)-3-Bromocamphor-8-sulfonic Acid Ammonium Salt
EINECSNumber (4)-1742
TSCA No
Recommended Storage Freezer