accessibility menu, dialog, popup

UserName

NH4H3CSOOOCH3OBr

CAS RN 55870-50-3

IUPAC Name: ammonium {3-bromo-1,7-dimethyl-2-oxobicyclo[2.2.1]heptan-7-yl}methanesulfonate
Synonyms: ammonium {3-bromo-1,7-dimethyl-2-oxobicyclo[2.2.1]heptan-7-yl}methanesulfonate
Molecular Weight (g/mol): 328.22
Molecular Formula: C10H18BrNO4S
InChi Key: GFBVBBRNPGPROZ-UHFFFAOYNA-N
SMILES: [NH4+].CC1(CS([O-])(=O)=O)C2CCC1(C)C(=O)C2Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results