Learn More
3-Bromobiphenyl, 99%
CAS: 2113-57-7 | C12H9Br | 233.11 g/mol
Supplier: Thermo Scientific Chemicals B2172614
Description
3-Bromobiphenyl is utilized as an electrochemical immunosensor based on poly(dopamine) coated gold nanocluster for brominated flame retardants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Bromobiphenyl is utilized as an electrochemical immunosensor based on poly(dopamine) coated gold nanocluster for brominated flame retardants.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
| 3-Bromobiphenyl | |
| 1.397 | |
| >110°C (230°F) | |
| 1.641 | |
| 25 g | |
| 3-bromobiphenyl, 3-bromo-1,1'-biphenyl, 1,1'-biphenyl, 3-bromo, 3-bromo-biphenyl, m-bromobiphenyl, biphenyl, 3-bromo, 3-bromodiphenyl, unii-o7i53k56s8, ccris 5889, 3-brombiphenyl | |
| USYQKCQEVBFJRP-UHFFFAOYSA-N | |
| 1-bromo-3-phenylbenzene | |
| 16449 | |
| 99% |
| 2113-57-7 | |
| 301°C | |
| C12H9Br | |
| MFCD00000082 | |
| 2043191 | |
| Immiscible with water. | |
| BrC1=CC=CC(=C1)C1=CC=CC=C1 | |
| 233.11 | |
| 233.12 |
Chemical Identifiers
| 2113-57-7 | |
| 233.11 | |
| USYQKCQEVBFJRP-UHFFFAOYSA-N | |
| 16449 | |
| BrC1=CC=CC(=C1)C1=CC=CC=C1 |
| C12H9Br | |
| MFCD00000082 | |
| 3-bromobiphenyl, 3-bromo-1,1'-biphenyl, 1,1'-biphenyl, 3-bromo, 3-bromo-biphenyl, m-bromobiphenyl, biphenyl, 3-bromo, 3-bromodiphenyl, unii-o7i53k56s8, ccris 5889, 3-brombiphenyl | |
| 1-bromo-3-phenylbenzene |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 218-304-9
RTECSNumber : DV1750000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only